Assay Details
Target Gene Details
Entrez Gene ID: | 13848 |
Gene Name: | Eph receptor B6 |
Gene Aliases: |
Cekl, Mep |
Location: |
Chr.6:41605040-41620507 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ephb6 | NM_001146351.1 | 13 | 2643 | NP_001139823.1 |
NM_007680.4 | 13 | 2639 | NP_031706.3 | |
AK076442.1 | 13 | 2466 | ||
AK087423.1 | 13 | 2554 | BAC39868.1 | |
AK163823.1 | 4 | 361 | BAE37507.1 | |
BC031924.1 | 12 | 2251 | AAH31924.1 | |
L77867.1 | 13 | 2333 | AAB51430.1 |
More Information
Set Membership: |
Intragenic Exonic |