Assay Details
Target Gene Details
Entrez Gene ID: | 65106 |
Gene Name: | ADP-ribosylation factor-like 6 interacting protein 5 |
Gene Aliases: |
5930404D22Rik, AV001879, Aip-5, Gtrap3-18, addiscin |
Location: |
Chr.6:97210792-97233315 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl6ip5 | NM_022992.2 | 1 | 137 | NP_075368.1 |
AF133912.1 | 1 | 135 | AAD33050.1 | |
AF265214.1 | 1 | 108 | AAL74056.1 | |
AF421860.1 | 1 | 143 | AAL77876.1 | |
AK005259.1 | 1 | 134 | BAB23912.1 | |
AK008519.1 | 1 | 117 | BAB25717.1 | |
AK076519.1 | 1 | 134 | BAC36376.1 | |
AK077826.1 | 1 | 144 | ||
AK088205.1 | 1 | 132 | BAC40209.1 | |
BC003897.1 | 1 | 137 | AAH03897.1 | |
D87211.1 | 1 | 180 | BAC24103.1 |
More Information
Set Membership: |
Intragenic Exonic |