Assay Details
Target Gene Details
Entrez Gene ID: | 77574 |
Gene Name: | TRPM8 channel-associated factor 1 |
Gene Aliases: |
2810407D09Rik, 3321401G04Rik, A230020K05Rik, AI848529, Fam115a |
Location: |
Chr.6:42667999-42710110 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcaf1 | NM_029930.2 | 1 | 66 | NP_084206.1 |
AK013025.1 | ||||
AK014379.1 | BAC25433.1 | |||
AK020704.1 | BAB32184.1 | |||
AK053509.1 | 1 | 67 | BAC35410.1 | |
AK076242.1 | ||||
AK083917.1 | BAC39061.1 | |||
AK132148.1 | BAE20997.1 | |||
AK148593.1 | ||||
AK173013.1 | BAD32291.1 | |||
BC031718.1 | 1 | 94 | AAH31718.1 |