Assay Details
Target Gene Details
Entrez Gene ID: | 52163 |
Gene Name: | calcium/calmodulin-dependent protein kinase I |
Gene Aliases: |
AI505105, CaMKIalpha, D6Ertd263e |
Location: |
Chr.6:113334124-113343922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Camk1 | NM_133926.2 | NP_598687.1 | ||
AK089742.1 | BAC40950.1 | |||
AK134936.1 | BAE22345.1 | |||
AK170188.1 | BAE41623.1 | |||
AK205870.1 | ||||
BC014825.1 | AAH14825.1 |
More Information
Set Membership: |
Intragenic Non-exonic |