Assay Details
Target Gene Details
Entrez Gene ID: | 321015 |
Gene Name: | RIKEN cDNA 5330439B14 gene |
Gene Aliases: |
- |
Location: |
Chr.6:142614595-142626462 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
5330439B14Rik | NR_037679.1 | 2 | 979 | |
AK019938.1 | 2 | 980 | BAB31925.1 | |
BC147438.1 | ||||
BC147456.1 |
Target Gene Details
Entrez Gene ID: | 20928 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 9 |
Gene Aliases: |
AI414027, AI449286, SUR2A, SUR2B, Sur2 |
Location: |
Chr.6:142587862-142702760 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcc9 | NM_001044720.1 | NP_001038185.1 | ||
NM_011511.2 | NP_035641.1 | |||
NM_021041.2 | NP_066378.1 | |||
NM_021042.2 | NP_066379.2 | |||
AF003531.1 | AAB58753.1 | |||
AK052291.1 | BAC34920.1 | |||
D86037.2 | BAA12969.2 | |||
D86038.2 | BAA12970.2 | |||
DQ991969.1 | ||||
DQ991970.1 | ||||
U97066.1 | AAB58701.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |