Assay Details
Target Gene Details
Entrez Gene ID: | 30956 |
Gene Name: | aminoadipate-semialdehyde synthase |
Gene Aliases: |
LKR, LKR/SDH, LOR, LOR/SDH, Lorsdh, SDH |
Location: |
Chr.6:23072173-23132986 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aass | NM_013930.4 | 3 | 421 | NP_038958.2 |
AJ224761.1 | 2 | 312 | CAA12114.1 | |
AK044323.1 | 3 | 402 | ||
AK098141.1 | 3 | 362 | ||
AK136216.1 | 3 | 370 | BAE22882.1 | |
AK149400.1 | 3 | 346 | BAE28852.1 | |
BC005420.1 | 3 | 407 | AAH05420.1 | |
BY275733.1 | ||||
CJ074697.1 |
More Information
Set Membership: |
Intragenic Exonic |