Assay Details
Target Gene Details
Entrez Gene ID: | 22697 |
Gene Name: | zinc finger and SCAN domain containing 21 |
Gene Aliases: |
AI326272, CTfin51, RU49, Zfp-38, Zfp38, Zipro1 |
Location: |
Chr.5:138116899-138134265 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zscan21 | NM_001044703.2 | NP_001038168.1 | ||
NM_001044704.2 | NP_001038169.1 | |||
NM_001044705.2 | NP_001038170.1 | |||
NM_011757.3 | NP_035887.2 | |||
AK146776.1 | ||||
AK146875.1 | BAE27498.1 | |||
BC010591.1 | AAH10591.1 | |||
BU513148.1 | ||||
BY034027.1 | ||||
BY293243.1 | ||||
BY309906.1 | ||||
CF915830.1 | ||||
D10630.1 | BAA01480.1 | |||
DV651309.1 | ||||
U41671.1 | AAB03786.1 | |||
X63747.1 | CAA45280.1 |
More Information
Set Membership: |
Intragenic Non-exonic |