Assay Details
Target Gene Details
Entrez Gene ID: | 75387 |
Gene Name: | sirtuin 4 |
Gene Aliases: |
4930596O17Rik |
Location: |
Chr.5:115478006-115490050 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirt4 |