Assay Details
Target Gene Details
Entrez Gene ID: | 269704 |
Gene Name: | zinc finger protein 664 |
Gene Aliases: |
AW060232, D930038J03Rik, ZFOC1, Znf664 |
Location: |
Chr.5:124862705-124888630 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp664 | NM_001081750.1 | NP_001075219.1 | ||
AK039671.1 | ||||
AK086580.1 | ||||
AK143192.1 | ||||
AK165974.1 |
More Information
Set Membership: |
Intragenic Non-exonic |