Assay Details
Target Gene Details
Entrez Gene ID: | 403178 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 1 |
Gene Aliases: |
A330045H12Rik, Gm576 |
Location: |
Chr.5:110099225-110105953 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcxd1 | NM_207279.2 | NP_997162.2 | ||
BC039627.1 | AAH39627.1 |