Assay Details
Target Gene Details
Entrez Gene ID: | 68770 |
Gene Name: | putative homeodomain transcription factor 2 |
Gene Aliases: |
1110054G21Rik, 9530062N20Rik, AI447096, C77923 |
Location: |
Chr.5:20758664-20882124 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phtf2 | NM_172992.3 | NP_766580.3 | ||
AK035536.1 | ||||
AK037021.1 | BAC29673.1 | |||
AK042382.1 | BAC31245.1 | |||
AK042790.1 | BAC31368.2 | |||
AK152951.1 | ||||
AK163376.1 | ||||
AW060751.1 | ||||
BC052841.1 | AAH52841.1 | |||
CB245749.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |