Assay Details
Target Gene Details
Entrez Gene ID: | 18778 |
Gene Name: | phospholipase A2, group IB, pancreas |
Gene Aliases: |
Pla2a, sPLA2IB |
Location: |
Chr.5:115466241-115474717 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g1b | NM_011107.1 | NP_035237.1 | ||
AF094610.1 | AAG27064.1 | |||
AF097637.1 | AAD19896.1 | |||
AF162712.1 | AAD45806.1 | |||
AF187852.1 | AAF44297.1 | |||
AK007730.1 | BAB25218.1 | |||
AK007797.1 | BAB25263.1 | |||
AK008331.1 | BAB25608.1 | |||
AK008664.1 | BAB25819.1 | |||
AK008668.1 | BAB25822.1 | |||
AK008841.1 | BAB25922.1 | |||
AK008934.1 | BAB25976.1 | |||
AK008936.1 | BAB25978.1 | |||
AK009314.1 | BAB26212.1 | |||
AK028104.1 | BAC25749.1 | |||
AK028134.1 | BAC25763.1 | |||
BC145908.1 | ||||
BC145910.1 |
More Information
Set Membership: |
Intragenic Non-exonic |