Assay Details
Target Gene Details
Entrez Gene ID: | 18778 |
Gene Name: | phospholipase A2, group IB, pancreas |
Gene Aliases: |
Pla2a, sPLA2IB |
Location: |
Chr.5:115466241-115474717 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g1b | NM_011107.1 | 2 | 99 | NP_035237.1 |
AF094610.1 | 2 | 92 | AAG27064.1 | |
AF097637.1 | 2 | 80 | AAD19896.1 | |
AF162712.1 | 2 | 99 | AAD45806.1 | |
AF187852.1 | 2 | 78 | AAF44297.1 | |
AK007730.1 | 2 | 93 | BAB25218.1 | |
AK007797.1 | 2 | 98 | BAB25263.1 | |
AK008331.1 | 2 | 95 | BAB25608.1 | |
AK008664.1 | 2 | 95 | BAB25819.1 | |
AK008668.1 | 2 | 95 | BAB25822.1 | |
AK008841.1 | 2 | 93 | BAB25922.1 | |
AK008934.1 | 2 | 99 | BAB25976.1 | |
AK008936.1 | 2 | 93 | BAB25978.1 | |
AK009314.1 | 2 | 92 | BAB26212.1 | |
AK019009.1 | 2 | 93 | ||
AK028104.1 | 2 | 96 | BAC25749.1 | |
AK028134.1 | 2 | 99 | BAC25763.1 | |
BC145908.1 | 2 | 99 | ||
BC145910.1 | 2 | 99 |
More Information
Set Membership: |
Intragenic Exonic |