Assay Details
Target Gene Details
Entrez Gene ID: | 231903 |
Gene Name: | ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5) decarboxylase |
Gene Aliases: |
EG231903, Prhoxnb |
Location: |
Chr.5:147314984-147322440 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Urad | NM_001039678.2 | NP_001034767.1 | ||
BC147779.1 | ||||
BC147790.1 | ||||
DQ225767.1 | ABB46374.1 |
More Information
Set Membership: |
Intragenic Non-exonic |