Assay Details
Target Gene Details
Entrez Gene ID: | 13483 |
Gene Name: | dipeptidylpeptidase 6 |
Gene Aliases: |
B930011P16Rik, D5Buc3, D5Buc4, D5Buc5, DPP VI, Dpp-6, Gm1377, In(5)6H-p, Peplb, Rw |
Location: |
Chr.5:26817357-27727500 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpp6 | NM_001136060.2 | 10 | 1269 | NP_001129532.1 |
NM_001198886.1 | 10 | 1353 | NP_001185815.1 | |
NM_010075.2 | 10 | 1604 | NP_034205.1 | |
NM_207282.3 | 10 | 1457 | NP_997165.2 | |
AF092506.1 | 7 | 617 | AAC97365.1 | |
AF092507.1 | 10 | 1612 | AAC97366.1 | |
AK031903.1 | 10 | 1229 | ||
AK033808.1 | 6 | 561 | ||
AK051708.1 | 11 | 1435 | ||
AK158964.1 | 10 | 1458 | BAE34745.1 | |
BC048383.1 | 10 | 1120 | AAH48383.1 | |
BC085154.1 | 10 | 1353 | AAH85154.1 |
More Information
Set Membership: |
Intragenic Exonic |