Assay Details
Target Gene Details
Entrez Gene ID: | 66383 |
Gene Name: | IscU iron-sulfur cluster scaffold homolog (E. coli) |
Gene Aliases: |
2310020H20Rik, AA407971, Nifu, Nifun |
Location: |
Chr.5:113772812-113778282 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Iscu | NM_025526.4 | NP_079802.1 | ||
AK009021.1 | BAB26031.1 | |||
AK009421.1 | ||||
AK039834.1 | BAC30464.1 | |||
AK083247.1 | BAC38830.1 | |||
AK157864.1 | ||||
BC028800.1 | AAH28800.1 | |||
BC048409.1 | AAH48409.1 | |||
BC096049.1 | AAH96049.1 |
More Information
Set Membership: |
Intragenic Non-exonic |