Assay Details
Target Gene Details
Entrez Gene ID: | 14581 |
Gene Name: | growth factor independent 1 |
Gene Aliases: |
AW495828, Gfi-1, Pal-1, Pal1 |
Location: |
Chr.5:107716655-107726031 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfi1 | NM_001267621.1 | NP_001254550.1 | ||
NM_010278.2 | NP_034408.1 | |||
AK089065.1 | BAC40731.1 | |||
BC130219.1 | 3 | 503 | ||
BC152328.1 | ||||
BU698904.1 | ||||
U58972.1 | AAC52959.1 | |||
U78312.1 | AAB36829.1 |
More Information
Set Membership: |
Intragenic Non-exonic |