Assay Details
Target Gene Details
Entrez Gene ID: | 16193 |
Gene Name: | interleukin 6 |
Gene Aliases: |
Il-6 |
Location: |
Chr.5:30013161-30019968 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il6 | NM_031168.1 | NP_112445.1 | ||
AK089780.1 | 4 | 481 | ||
AK150440.1 | BAE29562.1 | |||
AK152189.1 | BAE31020.1 | |||
BC132458.1 | ||||
BC138766.1 | ||||
DQ788722.1 | ||||
J03783.1 | AAA39301.1 | |||
X06203.1 | CAA29560.1 | |||
X54542.1 | CAA38411.1 |