Assay Details
Target Gene Details
Entrez Gene ID: | 20311 |
Gene Name: | chemokine (C-X-C motif) ligand 5 |
Gene Aliases: |
AMCF-II, ENA-78, GCP-2, LIX, Scyb5, Scyb6 |
Location: |
Chr.5:90759298-90761625 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl5 | NM_009141.3 | NP_033167.2 | ||
AK044397.1 | ||||
AK156787.1 | 1 | 692 | ||
BC024392.1 | AAH24392.1 | |||
BQ829929.1 | ||||
U27267.1 | AAC52238.1 |
More Information
Set Membership: |
Intragenic Non-exonic |