Assay Details
Target Gene Details
Entrez Gene ID: | 20310 |
Gene Name: | chemokine (C-X-C motif) ligand 2 |
Gene Aliases: |
CINC-2a, GROb, Gro2, MIP-2, MIP-2a, Mgsa-b, Mip2, Scyb, Scyb2 |
Location: |
Chr.5:90903899-90905938 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl2 | NM_009140.2 | NP_033166.1 | ||
AK137628.1 | BAE23436.1 | |||
AK150450.1 | BAE29571.1 | |||
AK155458.1 | 1 | 566 | ||
AK155874.1 | 3 | 359 | BAE33476.1 | |
AK155916.1 | BAE33502.1 | |||
AK157079.1 | BAE33956.1 | |||
BC119511.1 | ||||
BC119512.1 | ||||
X53798.1 | CAA37807.1 |
More Information
Set Membership: |
Intragenic Non-exonic |