Assay Details
Target Gene Details
Entrez Gene ID: | 14825 |
Gene Name: | chemokine (C-X-C motif) ligand 1 |
Gene Aliases: |
Fsp, Gro1, KC, Mgsa, N51, Scyb1, gro |
Location: |
Chr.5:90891245-90893121 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl1 | NM_008176.3 | NP_032202.1 | ||
AK140312.1 | 1 | 149 | ||
BC037997.1 | AAH37997.1 | |||
BC132502.1 | ||||
BC132504.1 | ||||
BY335927.1 | ||||
J04596.1 | AAA40131.1 |
More Information
Set Membership: |
Intragenic Non-exonic |