Assay Details
Target Gene Details
Entrez Gene ID: | 57914 |
Gene Name: | cytokine receptor-like factor 2 |
Gene Aliases: |
CRLM2, Ly114, Tpte2, Tslpr |
Location: |
Chr.5:109554709-109559017 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crlf2 | NM_001164735.1 | 4 | 571 | NP_001158207.1 |
NM_016715.4 | 4 | 571 | NP_057924.3 | |
AB031333.1 | 4 | 519 | BAA92159.1 | |
AB039945.1 | 4 | 467 | BAA92684.1 | |
AF201963.1 | 4 | 622 | AAF82189.1 | |
AF232936.1 | 4 | 535 | AAF81676.1 | |
AK149928.1 | 4 | 572 | ||
AK170466.1 | 1 | 433 | BAE41814.1 | |
BC016580.1 | 4 | 649 | ||
BC023788.1 | 4 | 559 | AAH23788.1 |
More Information
Set Membership: |
Intragenic Exonic |