Assay Details
Target Gene Details
Entrez Gene ID: | 65105 |
Gene Name: | ADP-ribosylation factor-like 6 interacting protein 4 |
Gene Aliases: |
AA408210, AA408365, Aip-4, Srp25 |
Location: |
Chr.5:124116108-124118195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arl6ip4 | NM_144509.2 | NP_653092.1 | ||
AB035383.1 | BAA94743.1 | |||
AK133934.1 | BAE21935.1 | |||
BC063748.1 | AAH63748.1 |