Assay Details
Target Gene Details
Entrez Gene ID: | 231871 |
Gene Name: | diacylglycerol lipase, beta |
Gene Aliases: |
E330036I19Rik |
Location: |
Chr.5:143464487-143504442 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Daglb | NM_144915.3 | NP_659164.2 | ||
AK086747.1 | BAC39734.1 | |||
AK087884.1 | BAC40033.1 | |||
AK136372.1 | BAE22951.1 | |||
BC016105.1 | AAH16105.2 |
More Information
Set Membership: |
Intragenic Non-exonic |