Assay Details
Target Gene Details
Entrez Gene ID: | 231871 |
Gene Name: | diacylglycerol lipase, beta |
Gene Aliases: |
E330036I19Rik |
Location: |
Chr.5:143464487-143504442 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Daglb | NM_144915.3 | 2 | 363 | NP_659164.2 |
AK086747.1 | 2 | 273 | BAC39734.1 | |
AK087884.1 | 2 | 218 | BAC40033.1 | |
AK135554.1 | 2 | 243 | ||
AK136372.1 | 2 | 221 | BAE22951.1 | |
BC016105.1 | 2 | 259 | AAH16105.2 | |
BY314231.1 |
More Information
Set Membership: |
Intragenic Exonic |