Assay Details
Target Gene Details
Entrez Gene ID: | 73471 |
Gene Name: | RIKEN cDNA 1700066N21 gene |
Gene Aliases: |
- |
Location: |
Chr.5:87908588-87979351 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700066N21Rik | NR_045924.1 | |||
AK006908.1 |
Target Gene Details
Entrez Gene ID: | 12994 |
Gene Name: | casein kappa |
Gene Aliases: |
AW208918, CSN10, Csnk |
Location: |
Chr.5:87925575-87932545 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csn3 | NM_007786.4 | NP_031812.2 | ||
AK021350.1 | BAB32382.1 | |||
AK085710.1 | BAC39516.1 | |||
AK142607.1 | BAE25129.1 | |||
AK142673.1 | BAE25155.1 | |||
AK164788.1 | BAE37916.1 | |||
BC004601.1 | AAH04601.1 | |||
BC005653.1 | AAH05653.1 | |||
BC049949.1 | AAH49949.1 | |||
BC050269.1 | AAH50269.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |