Assay Details
Target Gene Details
Entrez Gene ID: | 74114 |
Gene Name: | carnitine O-octanoyltransferase |
Gene Aliases: |
1200003H03Rik |
Location: |
Chr.5:8966023-8997146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crot | NM_023733.3 | NP_076222.1 | ||
AK004567.2 | BAB23378.1 | |||
BC006593.1 | AAH06593.1 | |||
BC012308.1 | AAH12308.1 |
More Information
Set Membership: |
Intragenic Non-exonic |