Assay Details
Target Gene Details
Entrez Gene ID: | 21942 |
Gene Name: | tumor necrosis factor receptor superfamily, member 9 |
Gene Aliases: |
4-1BB, A930040I11Rik, AA408498, AI325004, CDw137, Cd137, ILA, Ly63 |
Location: |
Chr.4:150920155-150946104 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnfrsf9 | NM_001077508.1 | NP_001070976.1 | ||
NM_001077509.1 | NP_001070977.1 | |||
NM_011612.2 | NP_035742.1 | |||
AK019885.1 | ||||
AK154625.1 | BAE32724.1 | |||
BC028507.1 | AAH28507.1 | |||
DQ832278.1 | ||||
DQ832279.1 |
More Information
Set Membership: |
Intragenic Non-exonic |