Assay Details
Target Gene Details
Entrez Gene ID: | 22275 |
Gene Name: | uroporphyrinogen decarboxylase |
Gene Aliases: |
AI323803, Uro-d |
Location: |
Chr.4:116990212-116994436 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Urod | NM_009478.3 | NP_033504.2 | ||
AK002844.1 | ||||
AK007470.1 | ||||
AK135090.1 | ||||
AK172426.1 | BAE43002.1 | |||
BC008109.1 | AAH08109.1 |
More Information
Set Membership: |
Intragenic Non-exonic |