Assay Details
Target Gene Details
Entrez Gene ID: | 14211 |
Gene Name: | structural maintenance of chromosomes 2 |
Gene Aliases: |
5730502P04Rik, AI255214, AW545314, CAP-E, CAPE, Fin16, SMC-2, Smc2l1 |
Location: |
Chr.4:52439221-52488365 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc2 | NM_001301412.1 | 21 | 3133 | NP_001288341.1 |
NM_008017.4 | 21 | 3057 | NP_032043.3 | |
AJ534939.1 | 20 | 2888 | CAD59182.1 | |
AK145341.1 | 21 | 3087 | BAE26376.1 | |
BC071232.1 | 6 | 792 | AAH71232.1 | |
BC094380.1 | 21 | 3029 | AAH94380.1 | |
U42385.1 | 4 | 500 | AAB08867.1 |