Assay Details
Target Gene Details
Entrez Gene ID: | 545622 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 3 |
Gene Aliases: |
9530011I20Rik, PTP-H1, PTPCL |
Location: |
Chr.4:57190841-57339046 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptpn3 | NM_011207.2 | 2 | 248 | NP_035337.2 |
AK052928.1 | 2 | 249 | BAC35205.1 | |
BC115634.1 | 2 | 147 | ||
BC115635.1 | 2 | 145 |
More Information
Set Membership: |
Intragenic Exonic |