Assay Details
Target Gene Details
Entrez Gene ID: | 18786 |
Gene Name: | phospholipase A2, activating protein |
Gene Aliases: |
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3 |
Location: |
Chr.4:94565139-94603247 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plaa | NM_172695.2 | 2 | 425 | NP_766283.2 |
AK019070.1 | 2 | 430 | ||
AK075934.1 | 2 | 357 | BAC36064.1 | |
AK081641.1 | 2 | 315 | ||
AK162534.1 | 2 | 326 | ||
AK166122.1 | 2 | 330 | ||
BC139355.1 | 2 | 424 | ||
BC139356.1 | 2 | 424 | ||
BC139773.1 | 2 | 335 | ||
CA490654.1 | 2 | 462 |
More Information
Set Membership: |
Intragenic Exonic |