Assay Details
Target Gene Details
Entrez Gene ID: | 18781 |
Gene Name: | phospholipase A2, group IIC |
Gene Aliases: |
- |
Location: |
Chr.4:138725325-138744575 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g2c | NM_008868.3 | NP_032894.2 | ||
BC029347.1 | AAH29347.1 | |||
CA464794.1 | ||||
U18119.1 | AAC52936.1 |
Target Gene Details
Entrez Gene ID: | 212190 |
Gene Name: | UBX domain protein 10 |
Gene Aliases: |
5730509E04Rik, A830047D02, Ubxd3 |
Location: |
Chr.4:138709837-138738438 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubxn10 | NM_001285928.1 | NP_001272857.1 | ||
NM_001285929.1 | NP_001272858.1 | |||
AK046949.1 | BAC32924.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |