Assay Details
Target Gene Details
Entrez Gene ID: | 18780 |
Gene Name: | phospholipase A2, group IIA (platelets, synovial fluid) |
Gene Aliases: |
EF, Mom1, Pla2, sPLA2, sPla2-IIA |
Location: |
Chr.4:138831857-138835189 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g2a | NM_001082531.1 | NP_001076000.1 | ||
NR_002926.3 | ||||
AK131857.1 | ||||
BC045156.1 | AAH45156.1 | |||
U28244.1 | AAB06315.1 | |||
U32359.1 | AAB60494.1 | |||
X74266.1 | CAA52325.1 |
Target Gene Details
Entrez Gene ID: | 18784 |
Gene Name: | phospholipase A2, group V |
Gene Aliases: |
PLA2, sPLA2 |
Location: |
Chr.4:138799247-138863469 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g5 | NM_001122954.1 | NP_001116426.1 | ||
AK090021.1 | BAC41050.1 | |||
BY012884.1 | ||||
BY015217.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |