Assay Details
Target Gene Details
Entrez Gene ID: | 242481 |
Gene Name: | paralemmin 2 |
Gene Aliases: |
53-H12, B130066I24, R75368, pEN70 |
Location: |
Chr.4:57568248-57717146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Palm2 | NM_172868.3 | NP_766456.1 | ||
AK045344.1 | BAC32319.1 | |||
BC113156.1 | AAI13157.1 |