Assay Details
Target Gene Details
Entrez Gene ID: | 56309 |
Gene Name: | c-myc binding protein |
Gene Aliases: |
5730488M09Rik, AW552132, AW742590, Amy-1 |
Location: |
Chr.4:123905013-123912250 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mycbp | NM_019660.3 | NP_062634.2 | ||
AB015858.1 | BAA31257.1 | |||
AK033289.1 | ||||
AK077633.1 | BAC36916.1 | |||
AK077762.1 | BAC36997.1 | |||
AK077995.1 | BAC37092.1 | |||
AK145773.1 | BAE26643.1 | |||
BC028344.1 | AAH28344.1 | |||
BC048478.1 | AAH48478.1 |
More Information
Set Membership: |
Intragenic Non-exonic |