Assay Details
Target Gene Details
Entrez Gene ID: | 74761 |
Gene Name: | matrix-remodelling associated 8 |
Gene Aliases: |
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam |
Location: |
Chr.4:155839680-155844102 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mxra8 | NM_024263.4 | NP_077225.4 | ||
AB040488.1 | BAB68501.1 | |||
AB052097.1 | BAD05133.1 | |||
AK004732.1 | BAB23514.1 | |||
AK195482.1 | ||||
BC026438.1 | AAH26438.1 | |||
EU849596.1 | ||||
EU849597.1 |