Assay Details
Target Gene Details
Entrez Gene ID: | 18198 |
Gene Name: | muscle, skeletal, receptor tyrosine kinase |
Gene Aliases: |
Mdk4, Mlk, Nsk1, Nsk2, Nsk3 |
Location: |
Chr.4:58285640-58377848 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Musk | NM_001037127.2 | NP_001032204.2 | ||
NM_001037128.1 | NP_001032205.1 | |||
NM_001037129.1 | NP_001032206.1 | |||
NM_001037130.1 | NP_001032207.1 | |||
NM_001165996.1 | NP_001159468.1 | |||
NM_010944.2 | NP_035074.2 | |||
AB691772.1 | ||||
AY360453.1 | AAR12275.1 | |||
AY955296.1 | AAY40318.1 | |||
AY955297.1 | AAY40319.1 | |||
AY955298.1 | AAY40320.1 | |||
AY956403.1 | AAY40321.1 | |||
AY956404.1 | AAY40322.1 | |||
BC100342.1 | AAI00343.1 | |||
U37708.1 | AAA79203.1 | |||
U37709.1 | AAA79204.1 | |||
X86444.1 | CAA60165.1 |
More Information
Set Membership: |
Intragenic Non-exonic |