Assay Details
Target Gene Details
Entrez Gene ID: | 230828 |
Gene Name: | interleukin 22 receptor, alpha 1 |
Gene Aliases: |
9130219A07Rik, ENSMUSG00000073746, IL-22R, Il22r |
Location: |
Chr.4:135728159-135755383 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il22ra1 | NM_178257.2 | 4 | 545 | NP_839988.1 |
AK141289.1 | 4 | 546 | BAE24639.1 | |
AY103454.1 | 4 | 484 | AAM52222.1 | |
BC140972.1 | 4 | 551 | ||
BC140973.1 | 4 | 551 |
More Information
Set Membership: |
Intragenic Exonic |