Assay Details
Target Gene Details
Entrez Gene ID: | 230075 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6 |
Gene Aliases: |
Gm137 |
Location: |
Chr.4:40270663-40279368 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufb6 | NM_001033305.2 | NP_001028477.1 | ||
AI853615.1 | ||||
AK146762.1 | BAE27414.1 | |||
BC147062.1 | ||||
BC147063.1 |
More Information
Set Membership: |
Intragenic Non-exonic |