Assay Details
Target Gene Details
Entrez Gene ID: | 230145 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 |
Gene Aliases: |
9130206E10, A630062B03Rik |
Location: |
Chr.4:47091893-47123042 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Galnt12 | NM_172693.3 | 5 | 980 | NP_766281.1 |
AB175680.1 | 5 | 960 | BAD52068.1 | |
AK033638.1 | 5 | 981 | BAC28401.1 | |
AK042133.1 | 5 | 1013 | BAC31179.1 | |
AK043977.1 | 4 | 840 | ||
AK082014.1 | 5 | 1026 | ||
AK156751.1 | 5 | 1010 | BAE33836.1 | |
BC056425.1 | 5 | 991 | AAH56425.1 |
More Information
Set Membership: |
Intragenic Exonic |