Assay Details
Target Gene Details
Entrez Gene ID: | 67460 |
Gene Name: | 2,4-dienoyl CoA reductase 1, mitochondrial |
Gene Aliases: |
1200012F07Rik, Decr, Nadph |
Location: |
Chr.4:15917240-15945507 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Decr1 | NM_026172.3 | NP_080448.1 | ||
AK002756.1 | BAB22333.1 | |||
AK004725.2 | BAB23508.1 | |||
AK010473.1 | ||||
AK015692.1 | BAB29933.1 | |||
AK146003.1 | BAE26824.1 | |||
AK168490.1 | BAE40377.1 | |||
BC046972.1 | AAH46972.1 | |||
CT010405.1 | CAJ18611.1 |
More Information
Set Membership: |
Intragenic Non-exonic |