Assay Details
Target Gene Details
Entrez Gene ID: | 54709 |
Gene Name: | eukaryotic translation initiation factor 3, subunit I |
Gene Aliases: |
36kDa, D4Ertd632e, Eif3s2, Trip1 |
Location: |
Chr.4:129591974-129600648 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3i | NM_018799.2 | NP_061269.1 | ||
AF188297.1 | AAF01455.1 | |||
AF271072.1 | AAF76199.1 | |||
AK002896.1 | BAB22440.1 | |||
AK010781.1 | BAB27177.1 | |||
AK012375.1 | BAB28197.1 | |||
AK089340.1 | 2 | 1613 | BAC40847.1 | |
AK149955.1 | ||||
AK172208.1 | BAE42882.1 | |||
BC029625.1 | AAH29625.1 | |||
CJ079467.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |