Assay Details
Target Gene Details
Entrez Gene ID: | 230073 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 58 |
Gene Aliases: |
6430573D20Rik, C330021E21, RIG-I, RLR-1 |
Location: |
Chr.4:40203777-40239825 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx58 | NM_172689.3 | 1 | 78 | NP_766277.3 |
AK052871.1 | BAC35183.1 | |||
AK078287.1 | BAC37205.1 | |||
AK087261.1 | 1 | 79 | BAC39830.1 | |
AK128929.1 | ||||
AK155930.1 | BAE33508.1 | |||
BC027369.1 | 1 | 57 | ||
BC106095.1 | AAI06096.1 |