Assay Details
Target Gene Details
Entrez Gene ID: | 56226 |
Gene Name: | espin |
Gene Aliases: |
je |
Location: |
Chr.4:152120331-152152420 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Espn | NM_207687.2 | 9 | 1628 | NP_997570.1 |
NM_207688.2 | 4 | 657 | NP_997571.1 | |
NM_207689.2 | NP_997572.1 | |||
NM_207690.2 | NP_997573.1 | |||
NM_207691.2 | 4 | 657 | NP_997574.2 | |
AF239886.1 | 9 | 1628 | AAF98134.1 | |
AF540942.1 | 4 | 657 | AAO50326.1 | |
AF540943.1 | 4 | 657 | AAO50327.1 | |
AF540944.1 | AAO50328.1 | |||
AF540945.1 | AAO50329.1 | |||
AK145160.1 | 2 | 192 | BAE26267.1 | |
AK146701.1 | 2 | 157 | ||
AK165010.1 | 1 | 1166 | ||
BY739473.1 | 2 | 157 |
More Information
Set Membership: |
Intragenic Exonic |