Assay Details
Target Gene Details
Entrez Gene ID: | 72479 |
Gene Name: | hydroxysteroid dehydrogenase like 2 |
Gene Aliases: |
2610207I16Rik |
Location: |
Chr.4:59580458-59618694 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsdl2 | NM_001301367.1 | NP_001288296.1 | ||
NM_024255.3 | NP_077217.2 | |||
AJ293845.1 | CAC34587.1 | |||
AK011910.1 | BAB27910.1 | |||
AK085899.1 | BAC39563.1 | |||
AK136773.1 | BAE23126.1 | |||
AK144948.1 | ||||
AK145230.1 | BAE26313.1 | |||
AY725196.1 | AAU11505.1 | |||
BC002211.1 | AAH02211.1 |
More Information
Set Membership: |
Intragenic Non-exonic |