Assay Details
Target Gene Details
Entrez Gene ID: | 14622 |
Gene Name: | gap junction protein, beta 5 |
Gene Aliases: |
Cnx31.1, Cx31.1, Gjb-5 |
Location: |
Chr.4:127354809-127358164 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjb5 | NM_010291.3 | 1 | 72 | NP_034421.1 |
AK082917.1 | 1 | 73 | BAC38687.1 | |
M91236.1 | AAA37426.1 |
More Information
Set Membership: |
Intragenic Exonic |