Assay Details
Target Gene Details
Entrez Gene ID: | 230098 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 39 |
Gene Aliases: |
E130306D19Rik |
Location: |
Chr.4:43496139-43508769 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgef39 | NM_001013377.2 | NP_001013395.2 | ||
AK087493.1 | 4 | 1131 | ||
AK153880.1 | 6 | 699 | ||
AK163287.1 | BAE37276.1 | |||
BC080700.1 | AAH80700.1 |
More Information
Set Membership: |
Intragenic Non-exonic |