Assay Details
Target Gene Details
Entrez Gene ID: | 12307 |
Gene Name: | calbindin 1 |
Gene Aliases: |
Brain-2, CB, Calb, Calb-1 |
Location: |
Chr.4:15881264-15906709 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Calb1 | NM_009788.4 | NP_033918.1 | ||
AK002635.1 | BAB22249.1 | |||
AK005081.1 | BAB23805.1 | |||
AK005243.1 | BAB23900.1 | |||
AK028050.1 | BAC25721.1 | |||
AK028327.1 | BAC25882.1 | |||
AK048517.1 | BAC33354.1 | |||
AK077499.1 | BAC36831.1 | |||
AK078358.1 | BAC37236.1 | |||
BC016421.1 | AAH16421.1 | |||
M21531.1 | AAA39898.1 | |||
M23663.1 | AAA53198.1 |
More Information
Set Membership: |
Intragenic Non-exonic |