Assay Details
Target Gene Details
Entrez Gene ID: | 66264 |
Gene Name: | coiled coil domain containing 28B |
Gene Aliases: |
1110002E08Rik, 1810010N17Rik |
Location: |
Chr.4:129619274-129623908 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc28b | NM_025455.2 | NP_079731.2 | ||
AK003287.1 | BAB22692.1 | |||
AK007427.1 | BAB25032.1 | |||
AK009176.1 | BAC25244.1 | |||
AK028262.1 | BAC25848.1 | |||
AK031278.1 | 1 | 652 | ||
AK158016.1 | BAE34319.1 | |||
BC028873.1 | AAH28873.1 | |||
BQ031123.1 |
More Information
Set Membership: |
Intragenic Non-exonic |